Tributyltin hydride 97% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tri-n-butyltin hydride

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 250-1000 ppm

A13298-09 A13298-14 A13298-18 A13298-22 A13298-30
CAAAAA13298-09EA 69.99 CAD
CAAAAA13298-09 CAAAAA13298-14 CAAAAA13298-18 CAAAAA13298-22 CAAAAA13298-30
Tributyltin hydride 97% stabilised
Tributyltin hydride

stabilised with 250-1000ppm BHT Purity: 97% CAS No.: 688-73-3 Molecular Formula: C12H28Sn Formula Weight: 291.07 Storage Temperature: -30°C to -10°C Flash Point: 40°(104°F) Refractive Index: 1.4730 Odor: Odorless Melting Point: 0-1° Boiling Point: 68-74°/0.3mm Beilstein Reference: 3587329 UN No.: 2929 MDL No.: MFCD00009416

Formula: C₁₂H₂₈Sn
Boiling Pt: 68…74 °C/0.3 mm
Melting Pt: 0…1 °C
Density: 1.098
Flash Pt: 40 °C (104 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009416
CAS Number: 688-73-3
EINECS: 211-704-4
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR