Bis(tributyltin) oxide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13242-0E
A13242-22
A13242-36
CAAAAA13242-0EEA
1158.14
CAD
CAAAAA13242-0E
CAAAAA13242-22
CAAAAA13242-36
Bis(tributyltin) oxide 97%
Bis(tributyltin) oxide
Purity: 97%
CAS No.: 56-35-9
Molecular Formula: C24H54OSn2
Formula Weight: 596.07
Flash Point: 168°(334°F)
Refractive Index: 1.4865
Odor: Irritating
Melting Point: -45°
Boiling Point: 179-180°/2mm
Beilstein Reference: 745057
UN No.: 2788
MDL No.: MFCD00009418
Formula:
C₂₄H₅₄OSn₂ Boiling Pt: 179…180 °C/2 mm Melting Pt: –45 °C Density: 1.172 Flash Pt: 168 °C (334 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009418 CAS Number: 56-35-9 EINECS: 200-268-0 UN: 2788 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...