2'-Methylacetanilide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Acetotoluidide, o-Acetotoluidid, N-(o-Tolyl)acetamide, N-(2-Methylphenyl)acetamide

A13235-14 A13235-22
CAAAAA13235-14EA 66.04 CAD
CAAAAA13235-14 CAAAAA13235-22
2'-Methylacetanilide 98+%
2'-Methylacetanilide

Purity: 98+% CAS No.: 120-66-1 Molecular Formula: C9H11NO Formula Weight: 149.19 Melting Point: 109-112° Boiling Point: 296° Merck Reference: 14,74 MDL No.: MFCD00014961

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Boiling Pt: 296 °C
Melting Pt: 109…112 °C
Density: 1.160
Storage Temperature: Ambient
MDL Number: MFCD00014961
CAS Number: 120-66-1
EINECS: 204-414-4
Merck Index: 14,74

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR