Ethyl-3,4,5-rimethoxybenzoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 3,4,5-trimethoxybenzoate, 3,4,5-Trimethoxybenzoic acid ethyl ester

A13199-06 A13199-22
CAAAA13199-06EA 31.13 CAD
CAAAA13199-06 CAAAA13199-14 CAAAA13199-22
Ethyl-3,4,5-rimethoxybenzoate 98%
Ethyl 3,4,5-rimethoxybenzoate

Purity: 98% CAS No.: 6178-44-5 Molecular Formula: C12H16O5 Formula Weight: 240.26 MDL No.: MFCD00017525

Formula: C₁₂H₁₆O₅
MW: 240.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017525
CAS Number: 6178-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR