Barium carbonate 99%

Supplier: Thermo Scientific Chemicals
Warning

A13195-22 A13195-36
CAAAAA13195-14EA 63.53 CAD
CAAAAA13195-14 CAAAAA13195-22 CAAAAA13195-36
Barium carbonate 99%
Barium carbonate

Purity: 99% CAS No.: 513-77-9 Molecular Formula: CBaO3 Formula Weight: 197.35 Odor: Odorless Melting Point: 811° Boiling Point: dec. Merck Reference: 14,969 MDL No.: MFCD00003448

Formula: BaCO₃
Boiling Pt: Dec.
Melting Pt: 811 °C
Density: 4.43
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
ADR: 6.1,III
Merck Index: 14,969

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR