Sodium acetate, anhydrous 99%

Supplier: Thermo Scientific Chemicals

A13184-0B A13184-0I A13184-30
CAAAAA13184-0BEA 123.24 CAD
CAAAAA13184-0B CAAAAA13184-0I CAAAAA13184-30
Sodium acetate, anhydrous 99%
Sodium acetate
Formula: H₃CCOONa
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.528
Flash Pt: (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 14,8571

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR