Adipoyl dichloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Adipoyl chloride

A13168-14 A13168-22 A13168-36
CAAAAA13168-14EA 105.07 CAD
CAAAAA13168-14 CAAAAA13168-22 CAAAAA13168-36
Adipoyl dichloride 98%
Adipoyl dichloride

Purity: 98% CAS No.: 111-50-2 Molecular Formula: C6H8Cl2O2 Formula Weight: 183.03 Flash Point: 110°(230°F) Refractive Index: 1.4705 Odor: Acrid Boiling Point: 105-107°/2mm Beilstein Reference: 507709 UN No.: 3265 MDL No.: MFCD00000759

Formula: C₆H₈Cl₂O₂
MW: 183.03 g/mol
Boiling Pt: 125
Density: 1.255
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000759
CAS Number: 111-50-2
EINECS: 203-876-4
UN: 2922
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR