Tetramethylsilane 99.9%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TMS

A13148-14 A13148-22
CAAAAA13148-14EA 89.59 CAD
CAAAAA13148-14 CAAAAA13148-22
Tetramethylsilane 99.9%
Tetramethylsilane

Purity: 99.9% CAS No.: 75-76-3 Molecular Formula: C4H12Si Formula Weight: 88.23 Storage Temperature: 2°C to 8°C Flash Point: -27°(-16°F) Refractive Index: 1.3590 Odor: Mild Melting Point: -100° Boiling Point: 26-27° Beilstein Reference: 1696908 Merck Reference: 14,9229 UN No.: 2749 MDL No.: MFCD00008274

Formula: C₄H₁₂Si
Boiling Pt: 26…27 °C
Melting Pt: –100 °C
Density: 0.646
Flash Pt: –27 °C (–16 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008274
CAS Number: 75-76-3
EINECS: 200-899-1
UN: 2749
ADR: 3,I
Merck Index: 14,9229

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR