Resorcinol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13080-0B
A13080-0I
A13080-18
A13080-30
CAAAAA13080-0BEA
246.48
CAD
CAAAAA13080-0B
CAAAAA13080-0I
CAAAAA13080-18
CAAAAA13080-30
Resorcinol 99%
Resorcinol
Purity: 99%
CAS No.: 108-46-3
Molecular Formula: C6H6O2
Formula Weight: 110.11
Flash Point: 171°(339°F)
Odor: Phenol-like
pH: 4-6
Melting Point: 109-112°
Boiling Point: 280-281°
Beilstein Reference: 906905
Merck Reference: 14,8155
UN No.: 2876
MDL No.: MFCD00002269
Formula:
C₆H₆O₂ Boiling Pt: 280…281 °C Melting Pt: 109…112 °C Density: 1.272 Flash Pt: 171 °C (339 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002269 CAS Number: 108-46-3 EINECS: 203-585-2 UN: 2876 ADR: 6.1,III Merck Index: 14,8155 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...