Potassium hydrogen difluoride 98%

Supplier: Thermo Scientific Chemicals
Danger

A13031-0E A13031-22 A13031-36
CAAAAA13031-0EEA 398.16 CAD
CAAAAA13031-0E CAAAAA13031-22 CAAAAA13031-36
Potassium hydrogen difluoride 98%
Potassium hydrogen difluoride

Purity: 98% CAS No.: 7789-29-9 Molecular Formula: KHF2 Formula Weight: 78.10 Odor: Odorless pH: 1 Melting Point: 239° dec. Merck Reference: 14,7610 UN No.: 1811 MDL No.: MFCD00011403

Formula: HF₂K
Melting Pt: 239 °C (dec.)
Density: 2.370
Storage Temperature: Ambient
MDL Number: MFCD00011403
CAS Number: 7789-29-9
EINECS: 232-156-2
UN: 1811
ADR: 8,II
Merck Index: 14,7610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR