Methyl methacrylate 99% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methacrylic acid methyl ester
Stabilization:
Stabilised with hydroquinone (HQ)
A13030-AE
A13030-AP
CAAAAA13030-0FEA
763.76
CAD
CAAAAA13030-0F
CAAAAA13030-AE
CAAAAA13030-AP
Methyl methacrylate 99% stabilised
Methyl methacrylate (monomer)
Purity: 99%
CAS No.: 80-62-6
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 10°(50°F)
Refractive Index: 1.4150
Melting Point: -48°
Boiling Point: 100-101°
Beilstein Reference: 605459
Merck Reference: 14,5941
UN No.: 1247
MDL No.: MFCD00008587
Formula:
CH₂=C(CH₃)COOCH₃ MW: 100.12 g/mol Boiling Pt: 100 Melting Pt: -48 Density: 0.93 g/cm³ Flash Pt: 8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008587 CAS Number: 80-62-6 EINECS: 201-297-1 UN: 1247 ADR: 3 II REACH: 01-2119452498-28 Merck Index: 14,5941 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...