3,4-Dimethylphenylhydrazinium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13027-30
A13027-09
A13027-18
CAAAAA13027-30EA
1440.96
CAD
CAAAAA13027-30
CAAAA13027-09
CAAAA13027-18
3,4-Dimethylphenylhydrazinium chloride 98%
3,4-Dimethylphenylhydrazinium chloride
Purity: 98%
CAS No.: 60481-51-8
Molecular Formula: C8H12N2.HCl
Formula Weight: 172.66
Melting Point: ca 195° dec.
Beilstein Reference: 6096060
Formula:
C₈H₁₃ClN₂ Melting Pt: ca 195 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00052270 CAS Number: 60481-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...