Nickel acetate tetrahydrate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel diacetate tetrahydrate, Nickel (II) acetate tetrahydrate, Nickel di(acetate) tetrahydrate, Acetic acid nickel (II) salt tetrahydrate

A13026-0E A13026-22 A13026-36
CAAAAA13026-0EEA 431.34 CAD
CAAAAA13026-0E CAAAAA13026-22 CAAAAA13026-36
Nickel acetate tetrahydrate 98+%
Nickel acetate tetrahydrate

Purity: 98+% CAS No.: 6018-89-9 Molecular Formula: Ni(CH3COO)2.4H2O Formula Weight: 248.84 (176.79anhy) Odor: Light Melting Point: dec. Beilstein Reference: 3730764 Merck Reference: 14,6499 UN No.: 3077 MDL No.: MFCD00066973

Formula: C₄H₆NiO₄·4H₂O
MW: 248.84 g/mol
Boiling Pt: 16
Melting Pt: Dec.
Density: 1.744
Storage Temperature: Ambient
MDL Number: MFCD00066973
CAS Number: 6018-89-9
EINECS: 206-761-7
UN: 3077
ADR: 9 III
Merck Index: 14,6499

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR