Sodium fluoride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13019-0B
A13019-0I
A13019-30
CAAAAA13019-0BEA
141.41
CAD
CAAAAA13019-0B
CAAAAA13019-0I
CAAAAA13019-30
Sodium fluoride 99%
Sodium fluoride
Purity: 99%
CAS No.: 7681-49-4
Molecular Formula: FNa
Formula Weight: 41.99
Refractive Index: 1.336
Odor: Odorless
Boiling Point: 1704°
Merck Reference: 14,8618
UN No.: 1690
MDL No.: MFCD00003524
Formula:
NaF Boiling Pt: 1704 °C Melting Pt: 993 °C Density: 2.78 Storage Temperature: Ambient |
MDL Number:
MFCD00003524 CAS Number: 7681-49-4 EINECS: 231-667-8 UN: 1690 ADR: 6.1,III Merck Index: 14,8618 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...