Sulfanilamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Aminobenzenesulfonamide, 4-Aminobenzenesulphonamide

A13001-0E A13001-22 A13001-36
CAAAAA13001-0EEA 537.2 CAD
CAAAAA13001-0E CAAAAA13001-22 CAAAAA13001-36
Sulfanilamide 98%
Sulphanilamide

Sulfonamide antibacterial


Purity: 98% CAS No.: 63-74-1 Molecular Formula: C6H8N2O2S Formula Weight: 172.21 Melting Point: 163-168° Beilstein Reference: 511852 Merck Reference: 14,8925 MDL No.: MFCD00007939

Formula: H₂NC₆H₄SO₂NH₂
MW: 172.21 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 163…168 °C
Density: 1.08
Storage Temperature: Ambient
MDL Number: MFCD00007939
CAS Number: 63-74-1
EINECS: 200-563-4
Merck Index: 14,8925
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a76bb9ab6bdf23e9d846b24d4f57047dea1d5071&vid=f2f1c5e47a127cd33d3b4a2a31b1057c375456eb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR