1,2-Dimethoxyethane 99+% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimethyl glycol, Ethylene glycol dimethyl ether, DME
Stabilization:
≤ 0.01 % Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
A12986-0F
A12986-AE
A12986-AP
CAAAAA12986-0FEA
507.18
CAD
CAAAAA12986-0F
CAAAAA12986-AE
CAAAAA12986-AP
1,2-Dimethoxyethane 99+% stabilized
1,2-Dimethoxyethane
Purity: 99+%
CAS No.: 110-71-4
Molecular Formula: C4H10O2
Formula Weight: 90.12
Flash Point: -6°(21°F)
Refractive Index: 1.3800
Odor: Ether-like
Melting Point: -69°
Boiling Point: 84-86°
Beilstein Reference: 1209237
Merck Reference: 14,3224
UN No.: 2252
MDL No.: MFCD00008502
Formula:
C₄H₁₀O₂ MW: 90.12 g/mol Boiling Pt: 84…86 °C Melting Pt: –69 °C Density: 0.867 Flash Pt: –6 °C (21 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008502 CAS Number: 110-71-4 EINECS: 203-794-9 UN: 2252 ADR: 3 II Merck Index: 14,3224 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...