Cerium trichloride heptahydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

A12947-0B A12947-18 A12947-30
CAAAAA12947-0BEA 701.52 CAD
CAAAAA12947-0B CAAAAA12947-18 CAAAAA12947-30
Cerium trichloride heptahydrate 99%
Cerium trichloride heptahydrate

Purity: 99% CAS No.: 18618-55-8 Molecular Formula: CeCl3.7H2O Formula Weight: 372.60 (246.48anhy) Odor: Odorless Merck Reference: 14,1997 MDL No.: MFCD00149634

Formula: CeCl₃·7H₂O
Melting Pt: 848 °C
Density: 3.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149634
CAS Number: 18618-55-8
EINECS: 606-073-0
UN: 1759
ADR: 8,III
Merck Index: 14,1997

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR