Syringaldehyde 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12933-06
A12933-14
A12933-22
CAAAAA12933-06EA
49.45
CAD
CAAAAA12933-06
CAAAAA12933-14
CAAAAA12933-22
Syringaldehyde 98+%
Syringaldehyde
Purity: 98+%
CAS No.: 134-96-3
Molecular Formula: C9H10O4
Formula Weight: 182.18
Melting Point: 110-113°
Boiling Point: 192-193°/14mm
Merck Reference: 14,9015
MDL No.: MFCD00006943
Formula:
C₉H₁₀O₄ Boiling Pt: 192…193 °C/14 mm Melting Pt: 110…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006943 CAS Number: 134-96-3 EINECS: 205-167-5 Merck Index: 14,9015 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...