2-(3-Chloro-4-fluorophenyl)indole 98%

Supplier: Thermo Scientific Chemicals
Warning

A12920-03 A12920-06
CAAAA12920-03EA 96.38 CAD
CAAAA12920-03 CAAAA12920-06
2-(3-Chloro-4-fluorophenyl)indole 98%
2-(3-Chloro-4-fluorophenyl)indole

Purity: 98% CAS No.: 1868-88-8 Molecular Formula: C14H9ClFN Formula Weight: 245.69 Melting Point: 170-173° MDL No.: MFCD00068179

Formula: C₁₄H₉ClFN
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00068179
CAS Number: 1868-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR