Zinc sulfate heptahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A12915-0E A12915-36
CAAAAA12915-0EEA 218.04 CAD
CAAAAA12915-0E CAAAAA12915-36
Zinc sulfate heptahydrate 98%
Zinc sulphate heptahydrate

Purity: 98% CAS No.: 7446-20-0 Molecular Formula: O4SZn.7H2O Formula Weight: 287.54 (161.43anhy) pH: 4.4-6 Melting Point: 100° UN No.: 3077 MDL No.: MFCD00149894

Formula: ZnSO₄·7H₂O
MW: 287.54 g/mol
Melting Pt: 100 °C
Density: 1.97
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9 III
Merck Index: 14,10159

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR