Zinc acetate dihydrate 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate

A12909-22 A12909-36 A12909-A4
CAAAAA12909-22EA 55.46 CAD
CAAAAA12909-22 CAAAAA12909-36 CAAAAA12909-A4
Zinc acetate dihydrate 97+%
Zinc acetate dihydrate

Purity: 97+% CAS No.: 5970-45-6 Molecular Formula: Zn(OOCCH3)2.2H2O Formula Weight: 219.50 (183.47anhy) Odor: Weak, characteristic pH: 6-7 Melting Point: 237° Beilstein Reference: 3732513 Merck Reference: 14,10128 UN No.: 3077 MDL No.: MFCD00066961

Formula: (H₃CCOO)₂Zn·2H₂O
Melting Pt: 237 °C
Density: 1.735
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 14,10128

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR