Barium chloride dihydrate 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12905-0B
A12905-0I
A12905-30
CAAAAA12905-0BEA
159.58
CAD
CAAAAA12905-0B
CAAAAA12905-0I
CAAAAA12905-30
Barium chloride dihydrate 99+%
Barium chloride dihydrate
Purity: 99+%
CAS No.: 10326-27-9
Molecular Formula: BaCl2.2H2O
Formula Weight: 244.28 (208.25anhy)
Odor: Odorless
Melting Point: 113° -2H2O
UN No.: 1564
MDL No.: MFCD00149154
Formula:
BaCl₂·2H₂O Boiling Pt: 1560 °C (1013 hPa) Melting Pt: 113 °C (–H₂O of crystall.) Density: 3.097 Storage Temperature: Ambient |
MDL Number:
MFCD00149154 CAS Number: 10326-27-9 EINECS: 233-788-1 UN: 1564 ADR: 6.1,III Merck Index: 14,971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...