Potassium hydrogen phthalate 99%

Supplier: Thermo Scientific Chemicals

A12895-0B A12895-0I
CAAAAA12895-0BEA 235.42 CAD
CAAAAA12895-0B CAAAAA12895-0I CAAAAA12895-36
Potassium hydrogen phthalate 99%
Potassium hydrogen phthalate

Purity: 99% CAS No.: 877-24-7 Molecular Formula: C8H5KO4 Formula Weight: 204.23 Odor: Odorless Melting Point: 297° dec. Beilstein Reference: 3637128 Merck Reference: 14,7612 MDL No.: MFCD00013070

Formula: HOOCC₆H₄COOK
Melting Pt: 297 °C (dec.)
Density: 1.636
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
EINECS: 212-889-4
Merck Index: 14,7612
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR