Ammonium cerium(IV) nitrate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate

A12882-0B A12882-18 A12882-30
CAAAAA12882-0BEA 573.54 CAD
CAAAAA12882-0B CAAAAA12882-18 CAAAAA12882-30
Ammonium cerium(IV) nitrate 98+%
Ammonium cerium (IV) nitrate

Purity: 98+% CAS No.: 16774-21-3 Molecular Formula: H8CeN8O18 Formula Weight: 548.23 Odor: Pungent Merck Reference: 14,1992 MDL No.: MFCD00151121

Formula: (NH₄)₂Ce(NO₃)₆
MW: 548.22 g/mol
Melting Pt: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
EINECS: 240-827-6
UN: 3085
ADR: 5.1 III
Merck Index: 14,1992

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR