Phenylselenol 90+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Benzeneselenol, Selenophenol
A12876-03
A12876-06
A12876-14
CAAAAA12876-03EA
106.02
CAD
CAAAAA12876-03
CAAAAA12876-06
CAAAAA12876-14
Phenylselenol 90+%
Phenylselenol
Purity: 90+%
CAS No.: 645-96-5
Molecular Formula: C6H6Se
Formula Weight: 157.07
Flash Point: 70°(158°F)
Refractive Index: 1.6120
Odor: Stench
Boiling Point: 71-72°/18mm
Beilstein Reference: 385715
UN No.: 3440
MDL No.: MFCD00000001
Formula:
C₆H₆Se MW: 157.07 g/mol Boiling Pt: 71 Density: 1.479 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000001 CAS Number: 645-96-5 EINECS: 211-457-2 UN: 3440 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...