2,4,6-Trichloropyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

A12842-06 A12842-14 A12842-22
CAAAA12842-06EA 42.03 CAD
CAAAA12842-06 CAAAA12842-14 CAAAA12842-22
2,4,6-Trichloropyrimidine 98%
2,4,6-Trichloropyrimidine

Purity: 98% CAS No.: 3764-01-0 Molecular Formula: C4HCl3N2 Formula Weight: 183.43 Flash Point: 110°(230°F) Refractive Index: 1.5700 Odor: Irritating Melting Point: 21-25° Boiling Point: 213-215° Beilstein Reference: 118284 UN No.: 1759 MDL No.: MFCD00006063

Formula: C₄HCl₃N₂
Boiling Pt: 213…215 °C
Melting Pt: 21…25 °C
Density: 1.595
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006063
CAS Number: 3764-01-0
EINECS: 223-183-0
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR