Methyl-4,5-dimethoxy-2-nitrobenzoate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl 6-nitroveratrate, 4,5-Dimethoxy-2-nitrobenzoic acid methyl ester, Methyl 4,5-dimethoxy-2-nitrobenzoate
A12836-18
A12836-30
A12836-36
CAAAAA12836-09EA
62.81
CAD
CAAAAA12836-09
CAAAAA12836-18
CAAAAA12836-30
CAAAA12836-36
Methyl-4,5-dimethoxy-2-nitrobenzoate 97%
Methyl 4,5-dimethoxy-2-nitrobenzoate
Purity: 97%
CAS No.: 26791-93-5
Molecular Formula: C10H11NO6
Formula Weight: 241.20
Melting Point: 142-144°
Beilstein Reference: 2381747
Formula:
C₁₀H₁₁NO₆ Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007239 CAS Number: 26791-93-5 EINECS: 248-008-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...