Magnesium fluoride hydrate ≥87% (dried basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12830-0E
A12830-36
CAAAAA12830-0EEA
255.96
CAD
CAAAAA12830-0E
CAAAAA12830-36
Magnesium fluoride hydrate ≥87% (dried basis)
Magnesium fluoride hydrate
CAS No.: 7783-40-6
Molecular Formula: MgF2.xH2O
Formula Weight: 62.30(anhy)
Odor: Odorless
Merck Reference: 14,5665
MDL No.: MFCD00011108
Formula:
F₂Mg·H₂O MW: 62.30 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00011108 CAS Number: 7783-40-6 EINECS: 231-995-1 Merck Index: 14,5665 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...