Thiourea 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12828-0C
A12828-0E
A12828-22
A12828-36
CAAAAA12828-0CEA
791.58
CAD
CAAAAA12828-0C
CAAAAA12828-0E
CAAAAA12828-22
CAAAAA12828-36
Thiourea 99%
Thiourea
Reagent for organic synthesis
Purity: 99%
CAS No.: 62-56-6
Molecular Formula: CH4N2S
Formula Weight: 76.12
Odor: Odorless
Melting Point: 172-178°
Beilstein Reference: 605327
Merck Reference: 14,9367
UN No.: 3077
MDL No.: MFCD00008067
Formula:
NH₂CSNH₂ Melting Pt: 172…178 °C Density: 1.405 Storage Temperature: Ambient |
MDL Number:
MFCD00008067 CAS Number: 62-56-6 EINECS: 200-543-5 UN: 3077 ADR: 9,III Merck Index: 14,9367 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...