Bromoethane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12804-0E
A12804-22
A12804-36
CAAAAA12804-0EEA
221.2
CAD
CAAAAA12804-0E
CAAAAA12804-22
CAAAAA12804-36
Bromoethane 98%
Bromoethane
Purity: 98%
CAS No.: 74-96-4
Molecular Formula: C2H5Br
Formula Weight: 108.97
Flash Point: -23°(-9°F)
Physical Form: Clear liquid
Density: 1.461
Refractive Index: 1.4235
Sensitivity: Light Sensitive
Appearance: Colorless
Odor: Ether-like
Melting Point: -119°
Boiling Point: 37-39°
Beilstein Reference: 1209224
Merck Reference: 14,3771
UN No.: 1891
MDL No.: MFCD00000232
Formula:
BrH₂CCH₃ Boiling Pt: 37…39 °C Melting Pt: –119 °C Density: 1.461 Flash Pt: –23 °C (–9 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000232 CAS Number: 74-96-4 EINECS: 200-825-8 UN: 1891 ADR: 6.1,II Merck Index: 14,3771 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...