Benzeneselenenyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylselenium chloride, Phenylselenenyl chloride, Chloroselenobenzene, Phenylselenyl chloride

A12751-06 A12751-14
CAAAAA12751-06EA 132.09 CAD
CAAAAA12751-06 CAAAAA12751-14
Benzeneselenenyl chloride 98%
Benzeneselenenyl chloride

Purity: 98% CAS No.: 5707-04-0 Molecular Formula: C6H5ClSe Formula Weight: 191.52 Odor: Stench Melting Point: 60-65° Boiling Point: 120°/20mm Beilstein Reference: 1237091 UN No.: 2928 MDL No.: MFCD00000478

Formula: (C₆H₅)SeCl
MW: 191.52 g/mol
Boiling Pt: 237
Melting Pt: 60…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00000478
CAS Number: 5707-04-0
EINECS: 227-196-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR