1,1,1,3,3,3-Hexafluoro-2-propanol 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP

A12747-06 A12747-14 A12747-22
CAAAAA12747-06EA 74.58 CAD
CAAAAA12747-06 CAAAAA12747-14 CAAAAA12747-22
1,1,1,3,3,3-Hexafluoro-2-propanol 99+%
1,1,1,3,3,3-Hexafluoro-2-propanol

Purity: 99+% CAS No.: 920-66-1 Molecular Formula: C3H2F6O Formula Weight: 168.04 Refractive Index: 1.2750 Melting Point: -4° Boiling Point: 59-60° Beilstein Reference: 1841007 UN No.: 1760 MDL No.: MFCD00011651

Formula: C₃H₂F₆O
MW: 168.04 g/mol
Boiling Pt: 59…60 °C
Melting Pt: –4 °C
Density: 1.619
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011651
CAS Number: 920-66-1
EINECS: 213-059-4
UN: 2922
ADR: 8,III
Merck Index: 13,09598

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR