Lead(IV) oxide 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead dioxide

A12742-0E A12742-36
CAAAAA12742-0EEA 698.36 CAD
CAAAAA12742-0E CAAAAA12742-36
Lead(IV) oxide 97%
Lead (IV) oxide

Purity: 97% CAS No.: 1309-60-0 Molecular Formula: PbO2 Formula Weight: 239.19 Odor: Odorless Melting Point: 290° UN No.: 1872 MDL No.: MFCD00011165

Formula: O₂Pb
MW: 239.2 g/mol
Melting Pt: 290
Density: 9.38
Storage Temperature: Ambient
MDL Number: MFCD00011165
CAS Number: 1309-60-0
EINECS: 215-174-5
UN: 1872
ADR: 5.1 III
Merck Index: 14,5407

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR