Barium hydroxide octahydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

A12714-0C A12714-0E A12714-36
CAAAAA12714-0CEA 886.38 CAD
CAAAAA12714-0C CAAAAA12714-0E CAAAAA12714-36
Barium hydroxide octahydrate 97%
Barium hydroxide octahydrate

Purity: 97% CAS No.: 12230-71-6 Molecular Formula: H2BaO2.8H2O Formula Weight: 315.48 (171.36anhy) Odor: Odorless Melting Point: 78° UN No.: 3262 MDL No.: MFCD00149152

Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Boiling Pt: 780
Melting Pt: 78 °C
Density: 2.18
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8 III
Merck Index: 14,977

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR