4-Aminobenzoic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Vitamin Bx, para-Aminobenzoic acid, PABA, Vitamin H1

A12673-0B A12673-18 A12673-30
CAAAAA12673-0BEA 229.1 CAD
CAAAAA12673-0B CAAAAA12673-18 CAAAAA12673-30
4-Aminobenzoic acid 99%
4-Aminobenzoic acid

Purity: 99% CAS No.: 150-13-0 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Melting Point: ca 187° dec. Beilstein Reference: 471605 Merck Reference: 14,423 MDL No.: MFCD00007894

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: ∼ 187 °C (dec.)
Density: 1.374
Storage Temperature: Ambient
MDL Number: MFCD00007894
CAS Number: 150-13-0
EINECS: 205-753-0
Merck Index: 14,423

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR