2-Ethylcaproic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Ethylhexanoic acid

A12644-0D A12644-0F A12644-AE A12644-AP
CAAAAA12644-0DEA 383.94 CAD
CAAAAA12644-0D CAAAAA12644-0F CAAAAA12644-AE CAAAAA12644-AP
2-Ethylcaproic acid 99%
2-Ethylcaproic acid

Purity: 99% CAS No.: 149-57-5 Molecular Formula: C8H16O2 Formula Weight: 144.21 Flash Point: 114°(237°F) Refractive Index: 1.4250 Melting Point: -59° Boiling Point: 226-229° Beilstein Reference: 1750468 MDL No.: MFCD00002675

Formula: C₈H₁₆O₂
Boiling Pt: 226…229 °C
Melting Pt: –59 °C
Density: 0.91
Flash Pt: 114 °C (237 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002675
CAS Number: 149-57-5
EINECS: 205-743-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR