Dichlorophen 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12640-14
A12640-22
A12640-36
CAAAAA12640-14EA
61.62
CAD
CAAAAA12640-14
CAAAAA12640-22
CAAAAA12640-36
Dichlorophen 95%
Dichlorophen
Purity: 95%
CAS No.: 97-23-4
Molecular Formula: C13H10Cl2O2
Formula Weight: 269.13
Melting Point: 172-176°
Beilstein Reference: 1884514
Merck Reference: 14,3071
UN No.: 3077
MDL No.: MFCD00002322
Formula:
C₁₃H₁₀Cl₂O₂ Boiling Pt: 389 °C (1013 hPa) Melting Pt: 172…176 °C Flash Pt: ≥ 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002322 CAS Number: 97-23-4 EINECS: 202-567-1 UN: 3077 ADR: 9,III Merck Index: 14,3071 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...