Trimethylsilyl isocyanate 94%

Supplier: Thermo Scientific Chemicals
Danger

A12633-09 A12633-18 A12633-30
CAAAA12633-09EA 167.48 CAD
CAAAA12633-09 CAAAA12633-18 CAAAA12633-30
Trimethylsilyl isocyanate 94%
Trimethylsilyl isocyanate

Purity: 94% CAS No.: 1118-02-1 Molecular Formula: C4H9NOSi Formula Weight: 115.21 Storage Temperature: -30°C to -10°C Flash Point: -2°(28°F) Refractive Index: 1.3960 Odor: Acrid Melting Point: -49° Boiling Point: 90-92° Beilstein Reference: 1744962 UN No.: 2478 MDL No.: MFCD00001993

Formula: C₄H₉NOSi
MW: 115.21 g/mol
Boiling Pt: 90…92 °C
Melting Pt: –49 °C
Density: 0.859
Flash Pt: -2 °C(28 °F)
Storage Temperature: Freezer
MDL Number: MFCD00001993
CAS Number: 1118-02-1
EINECS: 214-256-8
UN: 2478
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR