3-Methoxybenzyl chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12630-14
A12630-18
A12630-30
CAAAAA12630-14EA
154.52
CAD
CAAAAA12630-14
CAAAA12630-18
CAAAA12630-30
3-Methoxybenzyl chloride 98+%
3-Methoxybenzyl chloride
Purity: 98+%
CAS No.: 824-98-6
Molecular Formula: C8H9ClO
Formula Weight: 156.61
Storage Temperature: 2°C to 8°C
Flash Point: 101°(213°F)
Refractive Index: 1.5440
Boiling Point: 122-123°/12mm
Beilstein Reference: 636684
UN No.: 1760
MDL No.: MFCD00000907
Formula:
C₈H₉ClO Boiling Pt: 122…123 °C/12 mm Density: 1.078 Flash Pt: 101 °C (213 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000907 CAS Number: 824-98-6 EINECS: 212-541-1 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...