Tetrabutylammonium hydroxide 40% (w/w) in methanol

Supplier: Thermo Scientific Chemicals
Danger

A12626-0F A12626-AE A12626-AP
CAAAAA12626-0FEA 1818.58 CAD
CAAAAA12626-0F CAAAAA12626-AE CAAAAA12626-AP
Tetrabutylammonium hydroxide 40% (w/w) in methanol
Tetrabutylammonium hydroxide

CAS No.: 2052-49-5 Molecular Formula: [CH3(CH2)3]4NOH Formula Weight: 259.48 Flash Point: 18°(64°F) Refractive Index: 1.400 Odor: Alcohol-like Beilstein Reference: 3571228 UN No.: 2924 MDL No.: MFCD00009425

Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Density: 0.910
Flash Pt: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3286
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR