6-Aminobenzothiazole 98+%

Supplier: Thermo Scientific Chemicals
Warning

A12601-03 A12601-06
CAAAA12601-03EA 95.59 CAD
CAAAA12601-03 CAAAA12601-06 CAAAA12601-14
6-Aminobenzothiazole 98+%
6-Aminobenzothiazole

Purity: 98+% CAS No.: 533-30-2 Molecular Formula: C7H6N2S Formula Weight: 150.20 Melting Point: 86-90° Beilstein Reference: 116381 Merck Reference: 14,425 MDL No.: MFCD00015461

Formula: C₇H₆N₂S
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00015461
CAS Number: 533-30-2
EINECS: 208-559-4
Merck Index: 14,425

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR