Methyl-p-anisate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methoxybenzoic acid methyl ester, 4-Anisic acid methyl ester, p-Anisic acid methyl ester, Methyl 4-methoxybenzoate

A12598-14 A12598-22
CAAAA12598-14EA 51.03 CAD
CAAAA12598-14 CAAAA12598-22
Methyl-p-anisate 98+%
Methyl p-anisate

Purity: 98+% CAS No.: 121-98-2 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Odor: Characteristic Boiling Point: 244-245° Beilstein Reference: 2208571 MDL No.: MFCD00008437

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 244…245 °C
Melting Pt: 48…51 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008437
CAS Number: 121-98-2
EINECS: 204-513-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR