Bismuth(III) carbonate basic

Supplier: Thermo Scientific Chemicals

Synonyms: Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate

A12483-0B A12483-18 A12483-30
CAAAAA12483-0BEA 489.8 CAD
CAAAAA12483-0B CAAAAA12483-18 CAAAAA12483-30
Bismuth(III) carbonate basic
Bismuth (III) carbonate basic

CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 Formula Weight: 509.97 Merck Reference: 14,1282 MDL No.: MFCD00010877

Formula: (BiO)₂CO₃
MW: 509.97 g/mol
Density: 6.86
Storage Temperature: Ambient
MDL Number: MFCD00010877
CAS Number: 5892-10-4
EINECS: 227-567-9
Merck Index: 14,1282

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR