2-Vinylnaphthalene 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) ≤ 0.4 %
A12470-06
A12470-14
CAAAA12470-06EA
117.55
CAD
CAAAA12470-06
CAAAA12470-14
2-Vinylnaphthalene 97% stabilized
2-Vinylnaphthalene
Purity: 97%
CAS No.: 827-54-3
Molecular Formula: C12H10
Formula Weight: 154.21
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₂H₁₀ Boiling Pt: 128…131 °C/13 mm Melting Pt: 62…67 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00004125 CAS Number: 827-54-3 EINECS: 212-573-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...