Potassium hydrogen carbonate 99%
Supplier: Thermo Scientific Chemicals
A12429-0C
A12429-0E
A12429-36
CAAAAA12429-0CEA
638.32
CAD
CAAAAA12429-0C
CAAAAA12429-0E
CAAAAA12429-36
Potassium hydrogen carbonate 99%
Potassium hydrogen carbonate
Purity: 99%
CAS No.: 298-14-6
Molecular Formula: KHCO3
Formula Weight: 100.12
Odor: Odorless
Merck Reference: 14,7609
MDL No.: MFCD00011402
Formula:
KHCO₃ Melting Pt: 292 °C Density: 2.17 Storage Temperature: Ambient |
MDL Number:
MFCD00011402 CAS Number: 298-14-6 EINECS: 206-059-0 Merck Index: 14,7609 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...