Potassium hydrogen sulfate 97%

Supplier: Thermo Scientific Chemicals
Danger

A12428-0C A12428-0E A12428-22 A12428-36
CAAAAA12428-0CEA 688.88 CAD
CAAAAA12428-0C CAAAAA12428-0E CAAAAA12428-22 CAAAAA12428-36
Potassium hydrogen sulfate 97%
Potassium hydrogen sulphate

Purity: 97% CAS No.: 7646-93-7 Molecular Formula: HKO4S Formula Weight: 136.17 Odor: Odorless Melting Point: 195-197° dec. UN No.: 2509 MDL No.: MFCD00011404

Formula: KHSO₄
Melting Pt: 195…197 °C (dec.)
Density: 2.240
Storage Temperature: Ambient
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Merck Index: 14,7613

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR