2-(Benzyloxy)benzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12425-09
A12425-18
A12425-30
CAAAA12425-09EA
124.5
CAD
CAAAA12425-09
CAAAA12425-18
CAAAA12425-30
2-(Benzyloxy)benzoic acid 98%
2-(Benzyloxy)benzoic acid
Purity: 98%
CAS No.: 14389-86-7
Molecular Formula: C14H12O3
Formula Weight: 228.25
Melting Point: 76-78°
MDL No.: MFCD00051940
Formula:
C₁₄H₁₂O₃ Melting Pt: 76…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051940 CAS Number: 14389-86-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...