Sodium cyanate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12413-0E
A12413-22
A12413-36
CAAAAA12413-0EEA
366.56
CAD
CAAAAA12413-0E
CAAAAA12413-22
CAAAAA12413-36
Sodium cyanate 95%
Sodium cyanate
Purity: 95%
CAS No.: 917-61-3
Molecular Formula: NaOCN
Formula Weight: 65.01
Melting Point: 550°
Beilstein Reference: 3594963
Merck Reference: 14,8604
MDL No.: MFCD00011121
Formula:
CNNaO Melting Pt: 550 °C Density: 1.89 Storage Temperature: Ambient |
MDL Number:
MFCD00011121 CAS Number: 917-61-3 EINECS: 213-030-6 Merck Index: 14,8604 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...