4-Aminopyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

A12405-0B A12405-18 A12405-30
CAAAA12405-0BEA 1485.2 CAD
CAAAA12405-0B CAAAA12405-18 CAAAA12405-30
4-Aminopyridine 98%
4-Aminopyridine

Purity: 98% CAS No.: 504-24-5 Molecular Formula: C5H6N2 Formula Weight: 94.12 Flash Point: 164°(327°F) Odor: Unpleasant Boiling Point: 273-274° Beilstein Reference: 105782 Merck Reference: 14,3933 UN No.: 2671 MDL No.: MFCD00006439

Formula: C₅H₆N₂
Boiling Pt: 273…274 °C
Melting Pt: 157…161 °C
Density: 1.26
Flash Pt: 164 °C (327 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006439
CAS Number: 504-24-5
EINECS: 207-987-9
UN: 2671
ADR: 6.1,II
Merck Index: 14,3933

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR