Sodium tetrafluoroborate 97%

Supplier: Thermo Scientific Chemicals
Danger

A12393-22 A12393-36 A12393-A3
CAAAAA12393-22EA 50.56 CAD
CAAAAA12393-22 CAAAAA12393-36 CAAAAA12393-A3
Sodium tetrafluoroborate 97%
Sodium tetrafluoroborate

Purity: 97% CAS No.: 13755-29-8 Molecular Formula: BF4Na Formula Weight: 109.79 Melting Point: 384° dec. Merck Reference: 14,8617 MDL No.: MFCD00003515

Formula: BF₄Na
Melting Pt: 384 °C (dec.)
Density: 2.47
Storage Temperature: Ambient
MDL Number: MFCD00003515
CAS Number: 13755-29-8
EINECS: 237-340-6
UN: 1759
ADR: 8,III
Merck Index: 14,8617

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR